2-(Methylamino)butanenitrile
Supplier: Matrix Scientific
077219-500MG
077219-1G
077219-5G
102665-072EA
550.77
USD
102665-072
102665-074
102665-076
2-(Methylamino)butanenitrile
2-(Methylamino)butanenitrile
Hazard: Irritant
Formula:
C₅H₁₀N₂ Boiling Pt: 66°C/15 Torr Density: 0.886 |
MDL Number:
MFCD10695820 CAS Number: 106588-24-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...