3,3'-Dithiobis(4-aminobenzoic acid)

Supplier: Matrix Scientific

077205-500MG 077205-1G 077205-5G
102665-040EA 184.62 USD
102665-040 102665-042 102665-044
3,3'-Dithiobis(4-aminobenzoic acid)
3,3'-Dithiobis(4-aminobenzoic acid)

Hazard: Irritant

Formula: C₁₄H₁₂N₂O₄S₂
MDL Number: MFCD20441327
CAS Number: 102449-89-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR