3-(3-Chloroisoxazol-5-yl)propanamide

Supplier: Matrix Scientific

077196-500MG 077196-1G
102664-990EA 516.92 USD
102664-990 102664-992
3-(3-Chloroisoxazol-5-yl)propanamide
3-(3-Chloroisoxazol-5-yl)propanamide

Hazard: Irritant

Formula: C₆H₇ClN₂O₂
MDL Number: MFCD19691758
CAS Number: 1322604-59-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR