1-Benzyl-5-methyl-3-phenyl-1H-pyrazole-4-carboxylic acid

Supplier: Matrix Scientific

077195-500MG 077195-1G
102664-986EA 387.69 USD
102664-986 102664-988
1-Benzyl-5-methyl-3-phenyl-1H-pyrazole-4-carboxylic acid
1-Benzyl-5-methyl-3-phenyl-1H-pyrazole-4-carboxylic acid

Hazard: Irritant

Formula: C₁₈H₁₆N₂O₂
MDL Number: MFCD19691753
CAS Number: 1322605-00-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR