[1-Allyl-1-(tetrahydro-2H-pyran-4-yl)but-3-en-1-yl]amine
Supplier: Matrix Scientific
077193-500MG
077193-1G
077193-5G
102664-974EA
646.15
USD
102664-974
102664-976
102664-978
[1-Allyl-1-(tetrahydro-2H-pyran-4-yl)but-3-en-1-yl]amine
[1-Allyl-1-(tetrahydro-2H-pyran-4-yl)but-3-en-1-yl]amine
Hazard: Irritant
Formula:
C₁₂H₂₁NO |
MDL Number:
MFCD20441323 CAS Number: 1338495-06-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...