4-(2-Chlorobenzyl)tetrahydro-2H-pyran-4-carboxylic acid

Supplier: Matrix Scientific

077191-500MG 077191-1G 077191-5G
102664-962EA 516.92 USD
102664-962 102664-964 102664-966
4-(2-Chlorobenzyl)tetrahydro-2H-pyran-4-carboxylic acid
4-(2-Chlorobenzyl)tetrahydro-2H-pyran-4-carboxylic acid

Hazard: Irritant

Formula: C₁₃H₁₅ClO₃
MDL Number: MFCD17404695
CAS Number: 1274095-99-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR