4-(2-Bromobenzyl)tetrahydro-2H-pyran-4-carboxylic acid

Supplier: Matrix Scientific

077190-500MG 077190-1G 077190-5G
102664-956EA 516.92 USD
102664-956 102664-958 102664-960
4-(2-Bromobenzyl)tetrahydro-2H-pyran-4-carboxylic acid
4-(2-Bromobenzyl)tetrahydro-2H-pyran-4-carboxylic acid

Hazard: Irritant

Formula: C₁₃H₁₅BrO₃
MDL Number: MFCD17600244
CAS Number: 1338495-18-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR