4-(1,3-Benzodioxol-5-ylmethyl)tetrahydro-2H-pyran-4-carboxylic acid
Supplier: Matrix Scientific
077187-500MG
077187-1G
077187-5G
102664-938EA
516.92
USD
102664-938
102664-940
102664-942
4-(1,3-Benzodioxol-5-ylmethyl)tetrahydro-2H-pyran-4-carboxylic acid
4-(1,3-Benzodioxol-5-ylmethyl)tetrahydro-2H-pyran-4-carboxylic acid
Hazard: Irritant
Formula:
C₁₄H₁₆O₅ |
MDL Number:
MFCD20441321 CAS Number: 1338495-24-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...