4-Benzyltetrahydro-2H-pyran-4-carboxylic acid
Supplier: Matrix Scientific
077186-500MG
077186-1G
077186-5G
102664-932EA
516.92
USD
102664-932
102664-934
102664-936
4-Benzyltetrahydro-2H-pyran-4-carboxylic acid
4-Benzyltetrahydro-2H-pyran-4-carboxylic acid
Hazard: Irritant
Formula:
C₁₃H₁₆O₃ |
MDL Number:
MFCD17257048 CAS Number: 1338494-64-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...