2-(Methoxycarbonyl)cyclohexanecarboxylic acid

Supplier: Matrix Scientific

077184-500MG 077184-1G
102664-922EA 676.92 USD
102664-922 102664-924
2-(Methoxycarbonyl)cyclohexanecarboxylic acid
2-(Methoxycarbonyl)cyclohexanecarboxylic acid

Hazard: Irritant

Formula: C₉H₁₄O₄
MDL Number: MFCD00154581
CAS Number: 7719-08-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR