N-(1H-1,2,3-Benzotriazol-1-ylmethyl)-4-fluoroaniline

Supplier: Matrix Scientific

077183-500MG 077183-1G 077183-5G
102664-916EA 258.46 USD
102664-916 102664-918 102664-920
N-(1H-1,2,3-Benzotriazol-1-ylmethyl)-4-fluoroaniline
N-(1H-1,2,3-Benzotriazol-1-ylmethyl)-4-fluoroaniline

Hazard: Irritant

Formula: C₁₃H₁₁FN₄
MDL Number: MFCD00439366
CAS Number: 158719-41-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR