2-{[(4-Bromo-2-ethylphenyl)amino]methyl}-1H-isoindole-1,3(2H)-dione

Supplier: Matrix Scientific

077182-500MG 077182-1G 077182-5G
102664-910EA 258.46 USD
102664-910 102664-912 102664-914
2-{[(4-Bromo-2-ethylphenyl)amino]methyl}-1H-isoindole-1,3(2H)-dione
2-{[(4-Bromo-2-ethylphenyl)amino]methyl}-1H-isoindole-1,3(2H)-dione

Hazard: Irritant

Formula: C₁₇H₁₅BrN₂O₂
MDL Number: MFCD17069390
CAS Number: 1241674-55-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR