2-{[(4-Bromo-2-methylphenyl)amino]methyl}-1H-isoindole-1,3(2H)-dione
Supplier: Matrix Scientific
077181-500MG
077181-1G
102664-906EA
258.46
USD
102664-906
102664-908
2-{[(4-Bromo-2-methylphenyl)amino]methyl}-1H-isoindole-1,3(2H)-dione
2-{[(4-Bromo-2-methylphenyl)amino]methyl}-1H-isoindole-1,3(2H)-dione
Hazard: Irritant
Formula:
C₁₆H₁₃BrN₂O₂ |
MDL Number:
MFCD00087181 CAS Number: 116595-38-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...