2-({[3-(Trifluoromethyl)phenyl]amino}methyl)-3a,4,7,7a-tetrahydro-1H-4,7-methanoisoindole-1,3-dione

Supplier: Matrix Scientific

077178-500MG 077178-1G 077178-5G
102664-890EA 258.46 USD
102664-890 102664-892 102664-894
2-({[3-(Trifluoromethyl)phenyl]amino}methyl)-3a,4,7,7a-tetrahydro-1H-4,7-methanoisoindole-1,3-dione
2-({[3-(Trifluoromethyl)phenyl]amino}methyl)-3a,4,7,7a-tetrahydro-1H-4,7-methanoisoindole-1,3-dione

Hazard: Irritant

Formula: C₁₇H₁₅F₃N₂O₂
MDL Number: MFCD17069387
CAS Number: 1241674-84-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR