5-Phenylbicyclo[2.2.1]heptane-2,3-dicarboxylic acid

Supplier: Matrix Scientific

077175-500MG 077175-1G 077175-5G
102664-874EA 258.46 USD
102664-874 102664-876 102664-878
5-Phenylbicyclo[2.2.1]heptane-2,3-dicarboxylic acid
5-Phenylbicyclo[2.2.1]heptane-2,3-dicarboxylic acid

Hazard: Irritant

Formula: C₁₅H₁₆O₄
MDL Number: MFCD09267685
CAS Number: 1212145-57-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR