5-[(4-{[2-(4-Methoxyphenyl)-2-oxoethoxy]carbonyl}phenyl)amino]-5-oxopentanoic acid

Supplier: Matrix Scientific

077173-500MG 077173-1G 077173-5G
102664-862EA 161.54 USD
102664-862 102664-864 102664-866
5-[(4-{[2-(4-Methoxyphenyl)-2-oxoethoxy]carbonyl}phenyl)amino]-5-oxopentanoic acid
5-[(4-{[2-(4-Methoxyphenyl)-2-oxoethoxy]carbonyl}phenyl)amino]-5-oxopentanoic acid

Hazard: Irritant

Formula: C₂₁H₂₁NO₇
MW: 399.4 g/mol
MDL Number: MFCD05881777
CAS Number: 774557-91-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR