4-[(4-{[2-(4-Methoxyphenyl)-1-methyl-2-oxoethoxy]carbonyl}phenyl)amino]-4-oxobutanoic acid

Supplier: Matrix Scientific

077172-500MG 077172-1G 077172-5G
102664-856EA 161.54 USD
102664-856 102664-858 102664-860
4-[(4-{[2-(4-Methoxyphenyl)-1-methyl-2-oxoethoxy]carbonyl}phenyl)amino]-4-oxobutanoic acid
4-[(4-{[2-(4-Methoxyphenyl)-1-methyl-2-oxoethoxy]carbonyl}phenyl)amino]-4-oxobutanoic acid

Hazard: Irritant

Formula: C₂₁H₂₁NO₇
MDL Number: MFCD07050599
CAS Number: 906137-85-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR