4-[(4-{[2-(4-Nitrophenyl)-2-oxoethoxy]carbonyl}phenyl)amino]-4-oxobutanoic acid

Supplier: Matrix Scientific

077171-500MG 077171-1G 077171-5G
102664-850EA 161.54 USD
102664-850 102664-852 102664-854
4-[(4-{[2-(4-Nitrophenyl)-2-oxoethoxy]carbonyl}phenyl)amino]-4-oxobutanoic acid
4-[(4-{[2-(4-Nitrophenyl)-2-oxoethoxy]carbonyl}phenyl)amino]-4-oxobutanoic acid

Hazard: Irritant

Formula: C₁₉H₁₆N₂O₈
MDL Number: MFCD06756122
CAS Number: 774557-89-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR