2-{[(3-Chlorophenyl)amino]methyl}-1H-isoindole-1,3(2H)-dione

Supplier: Matrix Scientific

077170-500MG 077170-1G
102664-846EA 258.46 USD
102664-846 102664-848
2-{[(3-Chlorophenyl)amino]methyl}-1H-isoindole-1,3(2H)-dione
2-{[(3-Chlorophenyl)amino]methyl}-1H-isoindole-1,3(2H)-dione

Hazard: Irritant

Formula: C₁₅H₁₁ClN₂O₂
MDL Number: MFCD00178390
CAS Number: 57154-20-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR