2-{[(4-Chloro-2-methylphenyl)amino]methyl}-1H-isoindole-1,3(2H)-dione
Supplier: Matrix Scientific
077168-500MG
077168-1G
102664-836EA
258.46
USD
102664-836
102664-838
2-{[(4-Chloro-2-methylphenyl)amino]methyl}-1H-isoindole-1,3(2H)-dione
2-{[(4-Chloro-2-methylphenyl)amino]methyl}-1H-isoindole-1,3(2H)-dione
Hazard: Irritant
Formula:
C₁₆H₁₃ClN₂O₂ |
MDL Number:
MFCD00086926 CAS Number: 199803-03-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...