2-{[(3-Chloro-2-methylphenyl)amino]methyl}-1H-isoindole-1,3(2H)-dione

Supplier: Matrix Scientific

077167-500MG 077167-1G 077167-5G
102664-830EA 258.46 USD
102664-830 102664-832 102664-834
2-{[(3-Chloro-2-methylphenyl)amino]methyl}-1H-isoindole-1,3(2H)-dione
2-{[(3-Chloro-2-methylphenyl)amino]methyl}-1H-isoindole-1,3(2H)-dione

Hazard: Irritant

Formula: C₁₆H₁₃ClN₂O₂
MDL Number: MFCD00087009
CAS Number: 199803-02-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR