(4-Methoxyphenyl)(1-phenylethyl)amine

Supplier: Matrix Scientific

077163-500MG 077163-1G 077163-5G
102664-806EA 387.69 USD
102664-806 102664-808 102664-810
(4-Methoxyphenyl)(1-phenylethyl)amine
(4-Methoxyphenyl)(1-phenylethyl)amine

Hazard: Irritant

Formula: C₁₅H₁₇NO
Boiling Pt: 140…150 °C (0.006 Torr)
MDL Number: MFCD01126450
CAS Number: 2743-01-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR