2-{[(2-Methoxyphenyl)amino]methyl}hexahydro-4,6-ethenocyclopropa[f]isoindole-1,3(3aH)-dione
Supplier: Matrix Scientific
077161-500MG
077161-1G
077161-5G
102664-794EA
258.46
USD
102664-794
102664-796
102664-798
2-{[(2-Methoxyphenyl)amino]methyl}hexahydro-4,6-ethenocyclopropa[f]isoindole-1,3(3aH)-dione
2-{[(2-Methoxyphenyl)amino]methyl}hexahydro-4,6-ethenocyclopropa[f]isoindole-1,3(3aH)-dione
Hazard: Irritant
Formula:
C₁₉H₂₀N₂O₃ |
MDL Number:
MFCD03835822 CAS Number: 1005096-78-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...