2-{[(5-Chloro-2-methylphenyl)amino]methyl}hexahydro-4,6-ethenocyclopropa[f]isoindole-1,3(3aH)-dione

Supplier: Matrix Scientific

077160-500MG 077160-1G
102664-790EA 258.46 USD
102664-790 102664-792
2-{[(5-Chloro-2-methylphenyl)amino]methyl}hexahydro-4,6-ethenocyclopropa[f]isoindole-1,3(3aH)-dione
2-{[(5-Chloro-2-methylphenyl)amino]methyl}hexahydro-4,6-ethenocyclopropa[f]isoindole-1,3(3aH)-dione

Hazard: Irritant

Formula: C₁₉H₁₉ClN₂O₂
MDL Number: MFCD03836642
CAS Number: 1005096-12-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR