2-{[(3-Methylphenyl)amino]methyl}hexahydro-4,6-ethenocyclopropa[f]isoindole-1,3(3aH)-dione

Supplier: Matrix Scientific

077159-500MG 077159-1G
102664-786EA 258.46 USD
102664-786 102664-788
2-{[(3-Methylphenyl)amino]methyl}hexahydro-4,6-ethenocyclopropa[f]isoindole-1,3(3aH)-dione
2-{[(3-Methylphenyl)amino]methyl}hexahydro-4,6-ethenocyclopropa[f]isoindole-1,3(3aH)-dione

Hazard: Irritant

Formula: C₁₉H₂₀N₂O₂
MDL Number: MFCD03274160
CAS Number: 1005095-15-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR