2-{[(5-Chloro-2-methoxyphenyl)amino]methyl}-1H-isoindole-1,3(2H)-dione

Supplier: Matrix Scientific

077158-500MG 077158-1G 077158-5G
102664-780EA 258.46 USD
102664-780 102664-782 102664-784
2-{[(5-Chloro-2-methoxyphenyl)amino]methyl}-1H-isoindole-1,3(2H)-dione
2-{[(5-Chloro-2-methoxyphenyl)amino]methyl}-1H-isoindole-1,3(2H)-dione

Hazard: Irritant

Formula: C₁₆H₁₃ClN₂O₃
MDL Number: MFCD00106892
CAS Number: 859299-43-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR