2-(5-Chloro-2-methoxyphenyl)-1,3-dioxoisoindoline-5-carboxylic acid

Supplier: Matrix Scientific

077157-500MG 077157-1G 077157-5G
102664-774EA 161.54 USD
102664-774 102664-776 102664-778
2-(5-Chloro-2-methoxyphenyl)-1,3-dioxoisoindoline-5-carboxylic acid
2-(5-Chloro-2-methoxyphenyl)-1,3-dioxoisoindoline-5-carboxylic acid

Hazard: Irritant

Formula: C₁₆H₁₀ClNO₅
MDL Number: MFCD03348377
CAS Number: 690967-38-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR