2-{[(2-Ethylphenyl)amino]methyl}-1H-isoindole-1,3(2H)-dione

Supplier: Matrix Scientific

077156-500MG 077156-1G 077156-5G
102664-768EA 258.46 USD
102664-768 102664-770 102664-772
2-{[(2-Ethylphenyl)amino]methyl}-1H-isoindole-1,3(2H)-dione
2-{[(2-Ethylphenyl)amino]methyl}-1H-isoindole-1,3(2H)-dione

Hazard: Irritant

Formula: C₁₇H₁₆N₂O₂
MDL Number: MFCD00175062
CAS Number: 301223-08-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR