5-{[4-(4-tert-Butylphenoxy)phenyl]amino}-5-oxopentanoic acid

Supplier: Matrix Scientific

077155-500MG 077155-1G 077155-5G
102664-762EA 193.85 USD
102664-762 102664-764 102664-766
5-{[4-(4-tert-Butylphenoxy)phenyl]amino}-5-oxopentanoic acid
5-{[4-(4-tert-Butylphenoxy)phenyl]amino}-5-oxopentanoic acid

Hazard: Irritant

Formula: C₂₁H₂₅NO₄
MDL Number: MFCD02822549
CAS Number: 356089-70-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR