4-{[4-(4-tert-Butylphenoxy)phenyl]amino}-4-oxobutanoic acid
Supplier: Matrix Scientific
077154-500MG
077154-1G
077154-5G
102664-756EA
193.85
USD
102664-756
102664-758
102664-760
4-{[4-(4-tert-Butylphenoxy)phenyl]amino}-4-oxobutanoic acid
4-{[4-(4-tert-Butylphenoxy)phenyl]amino}-4-oxobutanoic acid
Hazard: Irritant
Formula:
C₂₀H₂₃NO₄ MW: 341.41 g/mol |
MDL Number:
MFCD02822520 CAS Number: 356089-44-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...