2-[4-(4-tert-Butylphenoxy)phenyl]-1,3-dioxoisoindoline-5-carboxylic acid

Supplier: Matrix Scientific

077153-500MG 077153-1G 077153-5G
102664-750EA 161.54 USD
102664-750 102664-752 102664-754
2-[4-(4-tert-Butylphenoxy)phenyl]-1,3-dioxoisoindoline-5-carboxylic acid
2-[4-(4-tert-Butylphenoxy)phenyl]-1,3-dioxoisoindoline-5-carboxylic acid

Hazard: Irritant

Formula: C₂₅H₂₁NO₅
MDL Number: MFCD02822518
CAS Number: 356087-74-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR