2-(5,6-Dibromo-1,3-dioxooctahydro-2H-4,7-methanoisoindol-2-yl)-3-phenylpropanoic acid

Supplier: Matrix Scientific

077146-500MG 077146-1G 077146-5G
102664-708EA 161.54 USD
102664-708 102664-710 102664-712
2-(5,6-Dibromo-1,3-dioxooctahydro-2H-4,7-methanoisoindol-2-yl)-3-phenylpropanoic acid
2-(5,6-Dibromo-1,3-dioxooctahydro-2H-4,7-methanoisoindol-2-yl)-3-phenylpropanoic acid

Hazard: Irritant

Formula: C₁₈H₁₇Br₂NO₄
MDL Number: MFCD02207457
CAS Number: 386702-34-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR