4-(5-Methyl-1,3-dioxooctahydro-2H-isoindol-2-yl)benzoic acid
Supplier: Matrix Scientific
077144-500MG
077144-1G
077144-5G
102664-696EA
258.46
USD
102664-696
102664-698
102664-700
4-(5-Methyl-1,3-dioxooctahydro-2H-isoindol-2-yl)benzoic acid
4-(5-Methyl-1,3-dioxooctahydro-2H-isoindol-2-yl)benzoic acid
Hazard: Irritant
Formula:
C₁₆H₁₇NO₄ MW: 287.32 g/mol |
MDL Number:
MFCD02026641 CAS Number: 345288-36-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...