2-(3-Chlorophenyl)-1,3-dioxoisoindoline-5-carboxylic acid

Supplier: Matrix Scientific

077143-500MG 077143-1G 077143-5G
102664-690EA 258.46 USD
102664-690 102664-692 102664-694
2-(3-Chlorophenyl)-1,3-dioxoisoindoline-5-carboxylic acid
2-(3-Chlorophenyl)-1,3-dioxoisoindoline-5-carboxylic acid

Hazard: Irritant

Formula: C₁₅H₈ClNO₄
MDL Number: MFCD00809584
CAS Number: 166096-47-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR