4-(Methylthio)-2-(4,5,6,7-tetrachloro-1,3-dioxo-1,3-dihydro-2H-isoindol-2-yl)butanoic acid

Supplier: Matrix Scientific

077142-500MG 077142-1G 077142-5G
102664-684EA 193.85 USD
102664-684 102664-686 102664-688
4-(Methylthio)-2-(4,5,6,7-tetrachloro-1,3-dioxo-1,3-dihydro-2H-isoindol-2-yl)butanoic acid
4-(Methylthio)-2-(4,5,6,7-tetrachloro-1,3-dioxo-1,3-dihydro-2H-isoindol-2-yl)butanoic acid

Hazard: Irritant

Formula: C₁₃H₉Cl₄NO₄S
MDL Number: MFCD00847218
CAS Number: 351436-89-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR