3-(5,6-Dibromo-1,3-dioxooctahydro-2H-4,7-methanoisoindol-2-yl)propanoic acid
Supplier: Matrix Scientific
077139-500MG
077139-1G
077139-5G
102664-666EA
193.85
USD
102664-666
102664-668
102664-670
3-(5,6-Dibromo-1,3-dioxooctahydro-2H-4,7-methanoisoindol-2-yl)propanoic acid
3-(5,6-Dibromo-1,3-dioxooctahydro-2H-4,7-methanoisoindol-2-yl)propanoic acid
Hazard: Irritant
Formula:
C₁₂H₁₃Br₂NO₄ |
MDL Number:
MFCD01823371 CAS Number: 386702-30-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...