2-[4-(2-Naphthyloxy)phenyl]-1,3-dioxoisoindoline-5-carboxylic acid

Supplier: Matrix Scientific

077138-500MG 077138-1G 077138-5G
102664-660EA 161.54 USD
102664-660 102664-662 102664-664
2-[4-(2-Naphthyloxy)phenyl]-1,3-dioxoisoindoline-5-carboxylic acid
2-[4-(2-Naphthyloxy)phenyl]-1,3-dioxoisoindoline-5-carboxylic acid

Hazard: Irritant

Formula: C₂₅H₁₅NO₅
MDL Number: MFCD01465963
CAS Number: 352671-43-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR