4-[(2,5-Dimethylphenyl)amino]-4-oxobutanoic acid

Supplier: Matrix Scientific

077133-500MG 077133-1G 077133-5G
102664-630EA 161.54 USD
102664-630 102664-632 102664-634
4-[(2,5-Dimethylphenyl)amino]-4-oxobutanoic acid
4-[(2,5-Dimethylphenyl)amino]-4-oxobutanoic acid

Hazard: Irritant

Formula: C₁₂H₁₅NO₃
MDL Number: MFCD00029828
CAS Number: 198276-05-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR