2-(3-Nitrophenyl)-2-oxoethyl-3-aminobenzoate
Supplier: Matrix Scientific
077132-500MG
077132-1G
077132-5G
102664-624EA
258.46
USD
102664-624
102664-626
102664-628
2-(3-Nitrophenyl)-2-oxoethyl-3-aminobenzoate
2-(3-Nitrophenyl)-2-oxoethyl-3-aminobenzoate
Hazard: Irritant
Formula:
C₁₅H₁₂N₂O₅ |
MDL Number:
MFCD01174811 CAS Number: 351896-55-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...