(5,6-Dibromo-1,3-dioxooctahydro-2H-4,7-methanoisoindol-2-yl)acetic acid

Supplier: Matrix Scientific

077127-500MG 077127-1G 077127-5G
102664-594EA 161.54 USD
102664-594 102664-596 102664-598
(5,6-Dibromo-1,3-dioxooctahydro-2H-4,7-methanoisoindol-2-yl)acetic acid
(5,6-Dibromo-1,3-dioxooctahydro-2H-4,7-methanoisoindol-2-yl)acetic acid

Hazard: Irritant

Formula: C₁₁H₁₁Br₂NO₄
MDL Number: MFCD01122738
CAS Number: 380428-86-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR