3-(5,6-Dibromo-1,3-dioxooctahydro-2H-4,7-methanoisoindol-2-yl)benzoic acid

Supplier: Matrix Scientific

077126-500MG 077126-1G 077126-5G
102664-588EA 161.54 USD
102664-588 102664-590 102664-592
3-(5,6-Dibromo-1,3-dioxooctahydro-2H-4,7-methanoisoindol-2-yl)benzoic acid
3-(5,6-Dibromo-1,3-dioxooctahydro-2H-4,7-methanoisoindol-2-yl)benzoic acid

Hazard: Irritant

Formula: C₁₆H₁₃Br₂NO₄
MDL Number: MFCD01122698
CAS Number: 101758-77-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR