4-(5,6-Dibromo-1,3-dioxooctahydro-2H-4,7-methanoisoindol-2-yl)benzoic acid

Supplier: Matrix Scientific

077125-500MG 077125-1G 077125-5G
102664-582EA 161.54 USD
102664-582 102664-584 102664-586
4-(5,6-Dibromo-1,3-dioxooctahydro-2H-4,7-methanoisoindol-2-yl)benzoic acid
4-(5,6-Dibromo-1,3-dioxooctahydro-2H-4,7-methanoisoindol-2-yl)benzoic acid

Hazard: Irritant

Formula: C₁₆H₁₃Br₂NO₄
MDL Number: MFCD01122697
CAS Number: 101690-99-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR