2-(4,5,6,7-Tetrachloro-1,3-dioxo-1,3-dihydro-2H-isoindol-2-yl)benzoic acid
Supplier: Matrix Scientific
077120-500MG
077120-1G
077120-5G
102664-552EA
161.54
USD
102664-552
102664-554
102664-556
2-(4,5,6,7-Tetrachloro-1,3-dioxo-1,3-dihydro-2H-isoindol-2-yl)benzoic acid
2-(4,5,6,7-Tetrachloro-1,3-dioxo-1,3-dihydro-2H-isoindol-2-yl)benzoic acid
Hazard: Irritant
Formula:
C₁₅H₅Cl₄NO₄ |
MDL Number:
MFCD01046595 CAS Number: 351998-35-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...