2-(2,3-Dichlorophenyl)-1,3-dioxoisoindoline-5-carboxylic acid

Supplier: Matrix Scientific

077116-500MG 077116-1G 077116-5G
102664-530EA 193.85 USD
102664-530 102664-532 102664-534
2-(2,3-Dichlorophenyl)-1,3-dioxoisoindoline-5-carboxylic acid
2-(2,3-Dichlorophenyl)-1,3-dioxoisoindoline-5-carboxylic acid

Hazard: Irritant

Formula: C₁₅H₇Cl₂NO₄
MDL Number: MFCD00981465
CAS Number: 313260-34-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR