1-(Anilinomethyl)pyrrolidine-2,5-dione

Supplier: Matrix Scientific

077114-500MG 077114-1G 077114-5G
102664-518EA 387.69 USD
102664-518 102664-520 102664-522
1-(Anilinomethyl)pyrrolidine-2,5-dione
1-(Anilinomethyl)pyrrolidine-2,5-dione

Hazard: Irritant

Formula: C₁₁H₁₂N₂O₂
MDL Number: MFCD00583508
CAS Number: 13314-99-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR