2-{[(3,4-Dichlorophenyl)amino]methyl}-1H-isoindole-1,3(2H)-dione

Supplier: Matrix Scientific

077113-500MG 077113-1G 077113-5G
102664-512EA 387.69 USD
102664-512 102664-514 102664-516
2-{[(3,4-Dichlorophenyl)amino]methyl}-1H-isoindole-1,3(2H)-dione
2-{[(3,4-Dichlorophenyl)amino]methyl}-1H-isoindole-1,3(2H)-dione

Hazard: Irritant

Formula: C₁₅H₁₀Cl₂N₂O₂
MDL Number: MFCD00419538
CAS Number: 293766-12-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR