2-(3,4-Dichlorophenyl)-1,3-dioxoisoindoline-5-carboxylic acid

Supplier: Matrix Scientific

077112-500MG 077112-1G 077112-5G
102664-506EA 193.85 USD
102664-506 102664-508 102664-510
2-(3,4-Dichlorophenyl)-1,3-dioxoisoindoline-5-carboxylic acid
2-(3,4-Dichlorophenyl)-1,3-dioxoisoindoline-5-carboxylic acid

Hazard: Irritant

Formula: C₁₅H₇Cl₂NO₄
MDL Number: MFCD00406014
CAS Number: 285552-82-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR