2-(Anilinomethyl)-1H-isoindole-1,3(2H)-dione
Supplier: Matrix Scientific
077109-500MG
077109-1G
077109-5G
102664-488EA
387.69
USD
102664-488
102664-490
102664-492
2-(Anilinomethyl)-1H-isoindole-1,3(2H)-dione
2-(Anilinomethyl)-1H-isoindole-1,3(2H)-dione
Hazard: Irritant
Formula:
C₁₅H₁₂N₂O₂ |
MDL Number:
MFCD00221821 CAS Number: 13314-96-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...