4-Amino-5-[2-(trifluoromethoxy)benzyl]-4H-1,2,4-triazole-3-thiol
Supplier: Matrix Scientific
077105-500MG
077105-1G
102664-468EA
387.69
USD
102664-468
102664-470
4-Amino-5-[2-(trifluoromethoxy)benzyl]-4H-1,2,4-triazole-3-thiol
4-Amino-5-[2-(trifluoromethoxy)benzyl]-4H-1,2,4-triazole-3-thiol
Hazard: Irritant
Formula:
C₁₀H₉F₃N₄OS |
MDL Number:
MFCD20441320 CAS Number: 1338495-05-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...