3-[(7-Methoxy-2-naphthyl)oxy]pyrazine-2-carboxylic acid
Supplier: Matrix Scientific
077099-500MG
077099-1G
077099-5G
102664-458EA
387.69
USD
102664-458
102664-460
102664-462
3-[(7-Methoxy-2-naphthyl)oxy]pyrazine-2-carboxylic acid
3-[(7-Methoxy-2-naphthyl)oxy]pyrazine-2-carboxylic acid
Hazard: Irritant
Formula:
C₁₆H₁₂N₂O₄ |
MDL Number:
MFCD16618458 CAS Number: 1228552-69-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...